missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Titanium Standard for ICP 1000 ppm 5% HCl/HF, Ricca Chemical

Catalog No. RV010322100
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010322100 100 mL
RV010322250 250 mL
RV010322500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010322100 Supplier Ricca Chemical Company Supplier No. RV010322100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7664-39-3 , 7647-01-0 , 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Packaging Natural LDPE Bottle
CAS Min % 92.30%, 4.90%, 0.49%
CAS Max % 96.06%, 5.10%, 0.51%
Specific Gravity 1
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity