missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tin Standard for ICP/MS 10 ppm in 1% HNO3/HF, Ricca Chemical

Catalog No. RV010623100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010623100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010623100 Supplier Ricca Chemical Company Supplier No. RV010623100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 ppm in 1% HNO3/HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS Min % 96.53%, 0.98%, 0.49%, 0.00%
CAS Max % 100%, 1.02%, 0.51%, 0.00%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity