missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tartrate - OOC(CHOH)2COO2- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Catalog No. p-7074157
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010945100 100 mL
RV010945250 250 mL
RV010945500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010945100 Supplier Ricca Chemical Company Supplier No. RV010945100C
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 87-69-4 , 7732-18-5
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Packaging Amber Glass Bottle
CAS Min % 97.90%, 0.10%
CAS Max % 100%, 0.10%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity