missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Sodium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$379.31 - $959.28

Identifiants chimiques

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
RV010345100
Afficher les documents
Ricca Chemical Company
RV010345100N
100 mL
chaque for $379.31
Il en reste null
 
RV010345250
Afficher les documents
Ricca Chemical Company
RV010345250N
250 mL
chaque for $673.97
Il en reste null
 
RV010345500
Afficher les documents
Ricca Chemical Company
RV010345500N
500 mL
chaque for $959.28
Il en reste null
 
Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Spécifications

Identifiants chimiques

7631-99-4
84.99
VWDWKYIASSYTQR-UHFFFAOYSA-N
CHEBI:63005
[Na+].[O-][N+]([O-])=O
NNaO3
MFCD00011119
24268
sodium nitrate

Spécifications

Natural LDPE Bottle
95.68%,1.96%,0.36%
1
NNaO3
Approximately 0°C
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate
100 mL
CHEBI:63005
7631-99-4,7697-37-2,7732-18-5
99.58%,2.04%,0.38%
Liquid
MFCD00011119
Standard
[Na+].[O-][N+]([O-])=O
84.99
24268
High Purity
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'