missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Silicon Standard for ICP/MS 100 ppm in H2O, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010553100N

Catalog No. RV010553100


Only null left
Explore available promotions

Description

Description

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Silicon ICP-MS Standard
16919-19-0 , 7732-18-5
100%,0.06%
Liquid
Colorless
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)
100 mL
High Purity
Natural LDPE Bottle
97.94%,0.06%
1
F6H8N2Si
Approximately 0°C
Miscible with water
Standard
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
178.153
28145

Chemical Identifiers

16919-19-0
178.153
28145
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
F6H8N2Si
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions