missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Silicon Standard for ICP 10 g/L in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$2024.00 - $2024.00
Chemical Identifiers
| CAS | 16919-19-0 |
|---|---|
| Molecular Formula | F6H8N2Si |
| Molecular Weight (g/mol) | 178.153 |
| InChI Key | ITHIMUMYFVCXSL-UHFFFAOYSA-P |
| PubChem CID | 28145 |
| IUPAC Name | diazanium;hexafluorosilicon(2-) |
| SMILES | [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010415500
|
Ricca Chemical Company
RV010415500N |
500 mL |
Each for $2,024.00
|
|
|||||
Description
- 10 g/L in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 16919-19-0 | |
| 178.153 | |
| 28145 | |
| [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
| F6H8N2Si | |
| ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
| diazanium;hexafluorosilicon(2-) |
Specifications
| Natural Poly Bottle | |
| 91.79%,6.21% | |
| 1 | |
| F6H8N2Si | |
| Standard | |
| [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F | |
| 178.153 | |
| 28145 |
| 16919-19-0 , 7732-18-5 | |
| 95.53%,6.47% | |
| Liquid | |
| Approximately 0°C | |
| ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
| diazanium;hexafluorosilicon(2-) | |
| 500 mL | |
| High Purity |