missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Silicate - SiO32- Standard for Ion Chromatography 1.000 +/- 0.002 g/l in NaOH 1%, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Facility
$384.53 - $748.22
Chemical Identifiers
| CAS | 6834-92-0 |
|---|---|
| Molecular Formula | Na2O3Si |
| Molecular Weight (g/mol) | 122.062 |
| InChI Key | NTHWMYGWWRZVTN-UHFFFAOYSA-N |
| PubChem CID | 23266 |
| ChEBI | CHEBI:60720 |
| IUPAC Name | disodium;dioxido(oxo)silane |
| SMILES | [O-][Si](=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010939100
|
Ricca Chemical Company
RV010939100N |
100 mL |
Each for $384.53
|
|
|||||
|
RV010939250
|
Ricca Chemical Company
RV010939250N |
250 mL |
Each for $523.59
|
|
|||||
|
RV010939500
|
Ricca Chemical Company
RV010939500N |
500 mL |
Each for $748.22
|
|
|||||
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 6834-92-0 | |
| 122.062 | |
| 23266 | |
| disodium;dioxido(oxo)silane |
| Na2O3Si | |
| NTHWMYGWWRZVTN-UHFFFAOYSA-N | |
| CHEBI:60720 | |
| [O-][Si](=O)[O-].[Na+].[Na+] |
Specifications
| Natural LDPE Bottle | |
| 97.83%,0.16%,0.01% | |
| 1.01 | |
| Na2O3Si | |
| NTHWMYGWWRZVTN-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)silane | |
| 100 mL | |
| CHEBI:60720 |