missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Silicate - SiO32- Standard for Ion Chromatography 1.000 +/- 0.002 g/l in NaOH 1%, Ricca Chemical

Catalog No. RV010939100
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010939100 100 mL
RV010939250 250 mL
RV010939500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010939100 Supplier Ricca Chemical Company Supplier No. RV010939100N

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 6834-92-0 , 7732-18-5
Molecular Formula Na2O3Si
Molecular Weight (g/mol) 122.062
InChI Key NTHWMYGWWRZVTN-UHFFFAOYSA-N
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.83%, 0.16%, 0.01%
CAS Max % 100%, 0.16%, 0.01%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity