missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
VeriSpec™ Phosphorus Standard for ICP/MS 100 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010540100N
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
7722-76-1,7732-18-5 | |
100 mL | |
100%,0.0408% | |
MFCD00003396 | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
1 |
Natural LDPE Bottle | |
97.9608%,0.0392% | |
H6NO4P | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
phosphoric acid amine | |
24402 | |
High Purity | |
Liquid |
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
CHEBI:62982 | |
N.OP(O)(O)=O |
H6NO4P | |
MFCD00003396 | |
24402 | |
phosphoric acid amine |