missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Perchlorate - ClO4- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Facility
$384.53 - $384.53
Chemical Identifiers
| CAS | 7601-89-0 |
|---|---|
| Molecular Formula | ClNaO4 |
| Molecular Weight (g/mol) | 122.436 |
| InChI Key | BAZAXWOYCMUHIX-UHFFFAOYSA-M |
| PubChem CID | 522606 |
| IUPAC Name | sodium;perchlorate |
| SMILES | [O-]Cl(=O)(=O)=O.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010934100
|
Ricca Chemical Company
RV010934100N |
100 mL |
Each for $384.53
|
|
|||||
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7601-89-0 | |
| 122.436 | |
| 522606 | |
| [O-]Cl(=O)(=O)=O.[Na+] |
| ClNaO4 | |
| BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
| sodium;perchlorate |
Specifications
| Natural LDPE Bottle | |
| 97.88%,0.12% | |
| 1.01 | |
| ClNaO4 | |
| BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
| sodium;perchlorate | |
| 100 mL | |
| High Purity |
| 7601-89-0,7732-18-5 | |
| 100%,0.12% | |
| Liquid | |
| Approximately 0°C | |
| [O-]Cl(=O)(=O)=O.[Na+] | |
| 122.436 | |
| 522606 |