missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Palladium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010542100N

Catalog No. RV010542100


Only null left
Explore available promotions

Description

Description

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Palladium ICP-MS Standard
10102-05-3 , 7697-37-2 , 7732-18-5
99.94%,2.04%,0.02%
Liquid
MFCD00011169
<2
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
GPNDARIEYHPYAY-UHFFFAOYSA-N
palladium(2+) dinitrate
100 mL
High Purity
Natural LDPE Bottle
96.02%,1.96%,0.02%
1
N2O6Pd
Colorless
Approximately 0°C
Miscible with water
Standard
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
230.43
24932

Chemical Identifiers

10102-05-3
230.43
GPNDARIEYHPYAY-UHFFFAOYSA-N
palladium(2+) dinitrate
N2O6Pd
MFCD00011169
24932
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions