missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Oxalate - C2O42- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Catalog No. RV010933500
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010933500 500 mL
RV010933100 100 mL
RV010933250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010933500 Supplier Ricca Chemical Company Supplier No. RV010933500N

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 1113-38-8 , 7732-18-5
Molecular Formula C2H8N2O4
Molecular Weight (g/mol) 124.096
InChI Key VBIXEXWLHSRNKB-UHFFFAOYSA-N
PubChem CID 14213
IUPAC Name diazanium;oxalate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.86%, 0.14%
CAS Max % 100%, 0.14%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 500 mL
Grade High Purity