missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Nitrilotriacetate - N(CH2COO)33- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Facility
$380.00 - $738.69
Chemical Identifiers
| CAS | 139-13-9 |
|---|---|
| Molecular Formula | C6H9NO6 |
| Molecular Weight (g/mol) | 191.14 |
| MDL Number | MFCD00004287 |
| InChI Key | MGFYIUFZLHCRTH-UHFFFAOYSA-N |
| PubChem CID | 8758 |
| ChEBI | CHEBI:44557 |
| IUPAC Name | 2-[bis(carboxymethyl)amino]acetic acid |
| SMILES | OC(=O)CN(CC(O)=O)CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010930100
|
Ricca Chemical Company
RV010930100C |
100 mL |
Each for $380.00
|
|
|||||
|
RV010930250
|
Ricca Chemical Company
RV010930250C |
250 mL |
Each for $517.16
|
|
|||||
|
RV010930500
|
Ricca Chemical Company
RV010930500C |
500 mL |
Each for $738.69
|
|
|||||
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 139-13-9 | |
| 191.14 | |
| MGFYIUFZLHCRTH-UHFFFAOYSA-N | |
| CHEBI:44557 | |
| OC(=O)CN(CC(O)=O)CC(O)=O |
| C6H9NO6 | |
| MFCD00004287 | |
| 8758 | |
| 2-[bis(carboxymethyl)amino]acetic acid |
Specifications
| Amber Glass Bottle | |
| 97.90%,0.10% | |
| 1.01 | |
| C6H9NO6 | |
| Approximately 0°C | |
| OC(=O)CN(CC(O)=O)CC(O)=O | |
| 191.14 | |
| 8758 | |
| High Purity |