missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Niobium (Nb) Standard for AAS 1000 ppm in 5% HNO3/1% HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$252.09 - $547.38
Chemical Identifiers
| CAS | 7783-68-8 |
|---|---|
| Molecular Formula | F5Nb |
| Molecular Weight (g/mol) | 187.90 |
| MDL Number | MFCD00011129 |
| InChI Key | AOLPZAHRYHXPLR-UHFFFAOYSA-I |
| IUPAC Name | niobium(5+) pentafluoride |
| SMILES | [F-].[F-].[F-].[F-].[F-].[Nb+5] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010035100
|
Ricca Chemical Company
RV010035100N |
100 mL |
N/A
|
|
|||||
|
RV010035500
|
Ricca Chemical Company
RV010035500N |
500 mL |
N/A
|
|
|||||
Description
- 1000 ppm in 5% HNO3/1% HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7783-68-8 | |
| 187.90 | |
| AOLPZAHRYHXPLR-UHFFFAOYSA-I | |
| [F-].[F-].[F-].[F-].[F-].[Nb+5] |
| F5Nb | |
| MFCD00011129 | |
| niobium(5+) pentafluoride |
Specifications
| Natural LDPE Bottle | |
| 91.92%,4.90%,0.98%,0.20% | |
| 1.01 | |
| F5Nb | |
| UN2922 | |
| AOLPZAHRYHXPLR-UHFFFAOYSA-I | |
| niobium(5+) pentafluoride | |
| 100 mL |
| 7783-68-8,7697-37-2,7664-39-3,7732-18-5 | |
| 95.68%,5.10%,1.02%,0.20% | |
| Liquid | |
| MFCD00011129 | |
| Approximately 0°C | |
| [F-].[F-].[F-].[F-].[F-].[Nb+5] | |
| 187.90 | |
| High Purity |