missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Neodymium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$467.57 - $1192.10
Chemical Identifiers
| CAS | 16454-60-7 |
|---|---|
| Molecular Formula | H12N3NdO15 |
| Molecular Weight (g/mol) | 438.344 |
| InChI Key | VQVDTKCSDUNYBO-UHFFFAOYSA-N |
| PubChem CID | 204494 |
| IUPAC Name | neodymium(3+);trinitrate;hexahydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010297100
|
Ricca Chemical Company
RV010297100N |
100 mL |
Each for $467.57
|
|
|||||
|
RV010297250
|
Ricca Chemical Company
RV010297250N |
250 mL |
Each for $833.83
|
|
|||||
|
RV010297500
|
Ricca Chemical Company
RV010297500N |
500 mL |
Each for $1,192.10
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 16454-60-7 | |
| 438.344 | |
| 204494 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] |
| H12N3NdO15 | |
| VQVDTKCSDUNYBO-UHFFFAOYSA-N | |
| neodymium(3+);trinitrate;hexahydrate |
Specifications
| Natural LDPE Bottle | |
| 95.75%,1.96%,0.29% | |
| 1 | |
| H12N3NdO15 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3] | |
| 438.344 | |
| 204494 |
| 16454-60-7 , 7697-37-2 , 16454-60-7 | |
| 99.65%,2.04%,0.31% | |
| Liquid | |
| Approximately 0°C | |
| VQVDTKCSDUNYBO-UHFFFAOYSA-N | |
| neodymium(3+);trinitrate;hexahydrate | |
| 100 mL | |
| High Purity |