missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Molybdenum (Mo) Standard for AAS 1000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$220.66 - $388.69
Chemical Identifiers
| CAS | 12027-67-7 |
|---|---|
| Molecular Formula | H24Mo7N6O24 |
| Molecular Weight (g/mol) | 1163.85 |
| MDL Number | MFCD00064633 |
| InChI Key | MISTUIVWPMGXFX-UHFFFAOYSA-T |
| PubChem CID | 71306766 |
| IUPAC Name | hexaammonium dodecaoxododecacyclo[7.5.1.13,5.13,13.15,7.17,9.01,4.01,6.01,8.02,13.011,14.011,15]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium) |
| SMILES | [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010079100
|
Ricca Chemical Company
RV010079100N |
100 mL |
Each for $220.66
|
|
|||||
|
RV010079500
|
Ricca Chemical Company
RV010079500N |
500 mL |
Each for $388.69
|
|
|||||
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 12027-67-7 | |
| 1163.85 | |
| MISTUIVWPMGXFX-UHFFFAOYSA-T | |
| hexaammonium dodecaoxododecacyclo[7.5.1.13,5.13,13.15,7.17,9.01,4.01,6.01,8.02,13.011,14.011,15]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium) |
| H24Mo7N6O24 | |
| MFCD00064633 | |
| 71306766 | |
| [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11 |
Specifications
| Natural LDPE Bottle | |
| 97.83%,0.17% | |
| 0.999 | |
| H24Mo7N6O24 | |
| Approximately 0°C | |
| [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11 | |
| 1163.85 | |
| 71306766 |
| 12027-67-7,7732-18-5 | |
| 100%,0.17% | |
| Liquid | |
| MFCD00064633 | |
| MISTUIVWPMGXFX-UHFFFAOYSA-T | |
| hexaammonium dodecaoxododecacyclo[7.5.1.13,5.13,13.15,7.17,9.01,4.01,6.01,8.02,13.011,14.011,15]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium) | |
| 100 mL | |
| High Purity |