missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Lithium Ionization Buffer for AAS 2% Li in 1% HNO3, Ricca Chemical

Catalog No. RV010142100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010142100 100 mL
RV010142500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010142100 Supplier Ricca Chemical Company Supplier No. RV010142100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7790-69-4 , 7697-37-2
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium(1+) nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 77.55%, 19.47%, 0.98%
CAS Max % 80.71%, 20.27%, 1.02%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity