missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Hydrogen Sulfite Standard for Ion Chromatography, 1000ppm in H2O, ISO 17025/Guide 34, Ricca Chemical
Formulated for high performance and reliable results
$599.84 - $599.84
Chemical Identifiers
| CAS | 7631-90-5 |
|---|---|
| Molecular Formula | NaO3S |
| Molecular Weight (g/mol) | 103.05 |
| MDL Number | MFCD00003530 |
| InChI Key | DWAQJAXMDSEUJJ-UHFFFAOYSA-L |
| PubChem CID | 23665763 |
| ChEBI | CHEBI:26709 |
| IUPAC Name | sodium sulfite |
| SMILES | [Na+].[O-]S([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010951500
|
Ricca Chemical Company
RV010951500N |
500 mL |
Each for $599.84
|
|
|||||
Description
- 1000ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7631-90-5 | |
| 103.05 | |
| DWAQJAXMDSEUJJ-UHFFFAOYSA-L | |
| CHEBI:26709 | |
| [Na+].[O-]S([O-])=O |
| NaO3S | |
| MFCD00003530 | |
| 23665763 | |
| sodium sulfite |
Specifications
| Bottle, Poly Natural | |
| 97.8726%,0.1274% | |
| 1.01 | |
| NaO3S | |
| DWAQJAXMDSEUJJ-UHFFFAOYSA-L | |
| sodium sulfite | |
| 500 mL | |
| CHEBI:26709 |