missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Hafnium Standard for ICP 1000 ppm in 2% HNO3/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$822.97 - $1734.47
Chemical Identifiers
| CAS | 12055-23-1 |
|---|---|
| Molecular Formula | HfO2 |
| Molecular Weight (g/mol) | 210.49 |
| MDL Number | MFCD00003565 |
| InChI Key | WIHZLLGSGQNAGK-UHFFFAOYSA-N |
| IUPAC Name | hafnium(4+) dioxidandiide |
| SMILES | [O--].[O--].[Hf+4] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010339100
|
Ricca Chemical Company
RV010339100N |
100 mL |
Each for $822.97
|
|
|||||
|
RV010339250
|
Ricca Chemical Company
RV010339250N |
250 mL |
Each for $1,169.13
|
|
|||||
|
RV010339500
|
Ricca Chemical Company
RV010339500N |
500 mL |
Each for $1,734.47
|
|
|||||
Description
- 1000 ppm in 2% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 12055-23-1 | |
| 210.49 | |
| WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
| [O--].[O--].[Hf+4] |
| HfO2 | |
| MFCD00003565 | |
| hafnium(4+) dioxidandiide |
Specifications
| Natural LDPE Bottle | |
| 94.94%,1.96%,0.98%,0.12% | |
| 1 | |
| HfO2 | |
| UN3287 | |
| Standard | |
| [O--].[O--].[Hf+4] | |
| 210.49 | |
| High Purity |
| 12055-23-1,7697-37-2,7664-39-3,7732-18-5 | |
| 98.82%,2.04%,1.02%,0.12% | |
| Liquid | |
| MFCD00003565 | |
| Approximately 0°C | |
| WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
| hafnium(4+) dioxidandiide | |
| 100 mL |