missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Formate - HCOO- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Facility

Supplier:  Ricca Chemical Company RV010916500N

 View more versions of this product

Catalog No. RV010916500


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
97.85%,0.15%
1.01
CHNaO2
HLBBKKJFGFRGMU-UHFFFAOYSA-M
sodium;formate
500 mL
CHEBI:62965
141-53-7 , 7732-18-5
100%,0.15%
Liquid
Approximately 0°C
C(=O)[O-].[Na+]
68.007
2723810
High Purity

Chemical Identifiers

141-53-7
68.007
2723810
sodium;formate
CHNaO2
HLBBKKJFGFRGMU-UHFFFAOYSA-M
CHEBI:62965
C(=O)[O-].[Na+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions