missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Europium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010278500N
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 95.82%,1.96%,0.22% | |
| 1 | |
| EuN3O9 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Eu+3] | |
| 337.976 | |
| 24983 |
| 10138-01-9,7697-37-2,7732-18-5 | |
| 99.74%,2.04%,0.22% | |
| Liquid | |
| Approximately 0°C | |
| GAGGCOKRLXYWIV-UHFFFAOYSA-N | |
| europium(3+);trinitrate | |
| 500 mL | |
| High Purity |
Chemical Identifiers
| 10138-01-9 | |
| 337.976 | |
| 24983 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Eu+3] |
| EuN3O9 | |
| GAGGCOKRLXYWIV-UHFFFAOYSA-N | |
| europium(3+);trinitrate |