missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Europium (Eu) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$569.59 - $1371.09
Chemical Identifiers
| CAS | 10138-01-9 |
|---|---|
| Molecular Formula | EuN3O9 |
| Molecular Weight (g/mol) | 337.976 |
| InChI Key | GAGGCOKRLXYWIV-UHFFFAOYSA-N |
| PubChem CID | 24983 |
| IUPAC Name | europium(3+);trinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Eu+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010018100
|
Ricca Chemical Company
RV010018100N |
100 mL |
Each for $569.59
|
|
|||||
|
RV010018500
|
Ricca Chemical Company
RV010018500N |
500 mL |
Each for $1,371.09
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10138-01-9 | |
| 337.976 | |
| 24983 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Eu+3] |
| EuN3O9 | |
| GAGGCOKRLXYWIV-UHFFFAOYSA-N | |
| europium(3+);trinitrate |
Specifications
| Natural LDPE Bottle | |
| 95.82%,1.96%,0.22% | |
| 1.01 | |
| EuN3O9 | |
| GAGGCOKRLXYWIV-UHFFFAOYSA-N | |
| europium(3+);trinitrate | |
| 100 mL | |
| High Purity |
| 10138-01-9,7697-37-2,7732-18-5 | |
| 99.74%,2.04%,0.22% | |
| Liquid | |
| Approximately 0°C | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Eu+3] | |
| 337.976 | |
| 24983 |