missing translation for 'onlineSavingsMsg'
Learn More

Verbascoside 97.0+%, TCI America™

Catalog No. V015310MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V015310MG 10 mg
V015350MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V015310MG Supplier TCI America Supplier No. V015310MG

Chemical Identifiers

CAS 61276-17-3
Molecular Formula C29H36O15
Molecular Weight (g/mol) 624.59
MDL Number MFCD00221751,MFCD00221751
InChI Key FBSKJMQYURKNSU-ZLSOWSIRSA-N
Synonym Acteoside, Kusaginin, 2-(3,4-Dihydroxyphenyl)ethyl 3-O-(6-Deoxy-alpha-L-mannopyranosyl)-4-O-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoyl]-beta-D-glucopyranoside
PubChem CID 45358104
IUPAC Name (2R,3R,4R,5R,6R)-6-[2-(3,4-dihydroxyphenyl)ethoxy]-5-hydroxy-2-(hydroxymethyl)-4-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-3-yl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCC3=CC=C(O)C(O)=C3)O[C@H](CO)[C@H]2OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 624.59
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Verbascoside
RTECSNumber LZ5786000
TSCA No