accessibility menu, dialog, popup

UserName

CAS RN 61276-17-3

IUPAC Name: (2R,3R,4R,5R,6R)-6-[2-(3,4-dihydroxyphenyl)ethoxy]-5-hydroxy-2-(hydroxymethyl)-4-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-3-yl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Synonyms: verbascoside acteoside kusaginin
Molecular Weight (g/mol): 624.59
Molecular Formula: C29H36O15
InChi Key: FBSKJMQYURKNSU-ZLSOWSIRSA-N
SMILES: C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCC3=CC=C(O)C(O)=C3)O[C@H](CO)[C@H]2OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results