missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Waste Funnels
VapLock™ Waste Funnels safely and securely capture waste and hazardous solvents
$831.29
Specifications
| For Use With (Application) | Solvent Safety |
|---|---|
| Type | Waste Funnel, 4 in. dia |
| Catalog Number | Mfr. No. | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
|
13311268
|
Cole-Parmer™
1202066 |
HDPE, Polypropylene | Container Ports with 1/2 in. NPT(F) |
Each for $831.29
|
|
||||
Description
- Reduces fumes and harmful vapor exhaust
- Easily connect to Justrite™ Centura safety containers
Warranty
None
Specifications
| Solvent Safety | |
| Waste Funnel, 4 in. dia |