Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
VapLock™ Solvent Waste Kits for Bottles and Carboys collect effluent from multiple sources while minimizing harmful solvent exhaust
Supplier: Cole-Parmer™ 1201868
Description
Various cap sizes available to fit your solvent containerWarranty
None
Chemical Identifiers
| Solvent-Compatible Containers | |
| 1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F) | |
| Solvent Safety | |
| Solvent Waste Kit |
Specifications
| Solvent-Compatible Containers | |
| Wako 38 mm | |
| Solvent Safety |
| Polypropylene | |
| 1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F) | |
| Solvent Waste Kit |