missing translation for 'onlineSavingsMsg'
Learn More

Vanillin 98.0+%, TCI America™

Supplier:  TCI America H0264500G

 View more versions of this product

Catalog No. H0264500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Vanillin
83°C
284°C
MFCD00006942,MFCD08702848
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
COC1=CC(C=O)=CC=C1O
152.15
CHEBI:18346
≥98.0% (GC,T)
121-33-5
White-Yellow
C8H8O3
500 g
MWOOGOJBHIARFG-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzaldehyde
1183
152.15
Crystalline Powder

Chemical Identifiers

121-33-5
152.15
MWOOGOJBHIARFG-UHFFFAOYSA-N
1183
4-hydroxy-3-methoxybenzaldehyde
C8H8O3
MFCD00006942,MFCD08702848
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
CHEBI:18346
COC1=CC(C=O)=CC=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1200

RTECSNumber : YW5775000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions