missing translation for 'onlineSavingsMsg'
Learn More

Vanillin 98.0+%, TCI America™

Catalog No. p-7130450
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H026425G 25 g
H0264500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H026425G Supplier TCI America Supplier No. H026425G
Only null left

Chemical Identifiers

CAS 121-33-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00006942,MFCD08702848
InChI Key MWOOGOJBHIARFG-UHFFFAOYSA-N
Synonym vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
PubChem CID 1183
ChEBI CHEBI:18346
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1O

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 284°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Vanillin
EINECSNumber (3)-1200
RTECSNumber YW5775000
TSCA Yes