missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Vandetanib
Description
Vandetanib is a potent VEGFR-2 inhibitor (IC50 = 0.04 μM), and inhibitor of EGFR (IC50 = 0.5 μM). Vandetanib potently inhibits (IC50 = 100 nM) RET oncoprotein activity in human thyroid cancer. Vandetanib displays anti-angiogenic properties in human nasopharyngeal carcinoma in mice model. Vandetanib is orally available and effective in vivo.
Specifications
Specifications
| Chemical Name or Material | N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl)methoxy]-4-quinazolinamine |
| CAS | 443913-73-3 |
| Quantity | 50 mg |
| Target | VEGF Receptor Inhibitors |
| Molecular Formula | C22H24BrFN4O2 |
| Purity | 0.98 |