missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Red |
| Physical Form | Solution |
| Quantity | 800 mL |
| Flash Point | −8°C |
| Packaging | AcroSeal glass Bottle |
| Specific Gravity | 1.312 |
| Density | 1.3120g/mL |
| Linear Formula | VCl·3THF |
| Formula Weight | 373.62 |
| Concentration or Composition (by Analyte or Components) | 0.40 to 0.60M |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS P Statement Highly flammable liquid and vapour. Suspected of causing cancer. Causes severe skin burns and eye damage. May cause respiratory irritation. May cause drowsiness or dizziness. May cause damage to organs |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
RUO – Research Use Only