CAS RN 19559-06-9
IUPAC Name:
oxolane; trichlorovanadium
Synonyms:
THF; trichlorovanadium
Molecular Weight (g/mol):
229.4
Molecular Formula:
C4H8Cl3OV
InChi Key:
WARDLUZTYLPMGJ-UHFFFAOYSA-K
SMILES:
Cl[V](Cl)Cl.C1CCOC1
1
–
1
of
1
results
1
Promotions Available
Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride, AcroSeal™
Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride, AcroSeal™
CAS: 19559-06-9 Molecular Formula: VCl3·C12H24O3 Molecular Weight (g/mol): 373.62 InChI Key: WARDLUZTYLPMGJ-UHFFFAOYSA-K Synonym: trichlorotris tetrahydrofuran vanadium,tetrahydrofuran; trichlorovanadium,vanadium iii chloride tetrahydrofuran adduct,vanadium iii chloride tetrahydrofuran complex PubChem CID: 71311352 IUPAC Name: oxolane;trichlorovanadium SMILES: C1CCOC1.Cl[V](Cl)Cl