Learn More
Valeraldehyde, 96%
CAS: 110-62-3 | C5H10O | 86.13 g/mol
$79.44 - $194.18
Chemical Identifiers
| CAS | 110-62-3 |
|---|---|
| Molecular Formula | C5H10O |
| Molecular Weight (g/mol) | 86.13 |
| MDL Number | MFCD00007026 |
| InChI Key | HGBOYTHUEUWSSQ-UHFFFAOYSA-N |
| Synonym | valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde |
| PubChem CID | 8063 |
| ChEBI | CHEBI:84069 |
| IUPAC Name | pentanal |
| SMILES | CCCCC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2138930
|
Thermo Scientific Chemicals
B2138930 |
250 g |
Each for $79.44
|
|
|||||
|
AAB213890B
|
Thermo Scientific Chemicals
B213890B |
1000 g |
Each for $194.18
|
|
|||||
Description
It finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.
Solubility
Soluble in water at 20°C, 13.5g/L. Soluble in alcohol.
Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 110-62-3 | |
| 86.13 | |
| HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
| 8063 | |
| pentanal |
| C5H10O | |
| MFCD00007026 | |
| valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
| CHEBI:84069 | |
| CCCCC=O |
Specifications
| 110-62-3 | |
| 0.81 | |
| 8°C (46°F) | |
| C5H10O | |
| CH3(CH2)3CHO | |
| 250 g | |
| 1616304 | |
| 14,9902 | |
| Soluble in water at 20°C,13.5g/L. Soluble in alcohol. | |
| CCCCC=O | |
| 86.13 | |
| CHEBI:84069 | |
| 97% |
| -92°C | |
| 102°C to 103°C | |
| Like Aldehyde | |
| 1.395 | |
| MFCD00007026 | |
| UN2058 | |
| Air Sensitive | |
| valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
| HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
| pentanal | |
| 8063 | |
| 86.13 | |
| Valeraldehyde |
Safety and Handling
GHS H Statement
H225-H318-H332-H335-H315
Highly flammable liquid and vapour.
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
H225-H317-H319-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VALERALDEHYDE
EINECSNumber : 203-784-4
RTECSNumber : YV3600000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only