missing translation for 'onlineSavingsMsg'
Learn More

Valeraldehyde, 96%

CAS: 110-62-3 | C5H10O | 86.13 g/mol

Supplier:  Thermo Scientific Chemicals B2138930

 View more versions of this product

Catalog No. AAB2138930


Only null left
Explore available promotions

Description

Description

It finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.

Solubility
Soluble in water at 20°C, 13.5g/L. Soluble in alcohol.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Valeraldehyde
-92°C
102°C to 103°C
Like Aldehyde
1.395
MFCD00007026
UN2058
Air Sensitive
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde
HGBOYTHUEUWSSQ-UHFFFAOYSA-N
pentanal
8063
86.13
110-62-3
0.81
8°C (46°F)
C5H10O
CH3(CH2)3CHO
250 g
1616304
14,9902
Soluble in water at 20°C,13.5g/L. Soluble in alcohol.
CCCCC=O
86.13
CHEBI:84069
97%

Chemical Identifiers

110-62-3
86.13
HGBOYTHUEUWSSQ-UHFFFAOYSA-N
8063
pentanal
C5H10O
MFCD00007026
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde
CHEBI:84069
CCCCC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H318-H332-H335-H315
Highly flammable liquid and vapour.
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c

H225-H317-H319-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VALERALDEHYDE

EINECSNumber : 203-784-4

RTECSNumber : YV3600000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only