accessibility menu, dialog, popup

UserName

H3CO

CAS RN 110-62-3

IUPAC Name: pentanal
Synonyms: pentanal fema number 3098 n-valeraldehyde (natural) valeraldehyde (substance) butyl formal valerianic aldehyde valeryl aldehyde
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: HGBOYTHUEUWSSQ-UHFFFAOYSA-N
SMILES: CCCCC=O
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)