Learn More
Cytiva V-Bottom 96-Well Collection Plate
Description
- Excellent fit to prepacked MultiTrap™ plates and PreDictor™ plates
- V-bottom for minimal loss of sample
- Collection of <500μL volumes of sample per well
Specifications
Specifications
| Product Type | V-botto m collection plate |
| Quantity | 5 Pack |
| Material | Polypropylene |
| Column Type | Column Accesory |
| For Use With (Application) | Collection of eluted samples in parallel from screening plates such as MultiTrap and PreDicto r plates |