missing translation for 'onlineSavingsMsg'
Learn More

United Scientific Supplies Utility Tray

Steam autoclavable, polypropylene molded trays.

Supplier:  United Scientific Supplies 81721

 View more versions of this product

Catalog No. S99413


Only null left

Description

Description

Steam autoclavable, polypropylene molded trays.

  • Ideal for sterilizing, drying or storage of labware.
  • Carrier tray has built-in handle for easy carrying of reagents or labware.
  • Standard Tray measures: 15 x 14 x 5 in. (38.1 x 35.6 x 12.7 cm)
  • Carrier Tray measures: 15 x 9.5 x 4.5 in. (38.1 x 24.1 x 11.4 cm)
Specifications

Specifications

15 x 14 x 5 in. (38 x 35.5 x 12.7 cm)
12.7 cm
Polypropylene
35.5 cm
5 in.
Autoclavable
White
38 cm
Utility Tray
15 in.
14 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions