missing translation for 'onlineSavingsMsg'
Learn More

United Scientific Supplies Utility Tray

Catalog No. p-3796007
Change view
Click to view available options
Type:
Carrier Tray
Utility Tray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S99414 Carrier Tray
S99413 Utility Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S99414 Supplier United Scientific Supplies Supplier No. EPD81731
Only null left

Steam autoclavable, polypropylene molded trays.

Steam autoclavable, polypropylene molded trays.

  • Ideal for sterilizing, drying or storage of labware.
  • Carrier tray has built-in handle for easy carrying of reagents or labware.
  • Standard Tray measures: 15 x 14 x 5 in. (38.1 x 35.6 x 12.7 cm)
  • Carrier Tray measures: 15 x 9.5 x 4.5 in. (38.1 x 24.1 x 11.4 cm)

Specifications

Dimensions (L x W x H) 15 x 9.5 x 4.5 in. (38 x 24 x 11.4 cm)
Color White
Height (Metric) 11.4 cm
Length (Metric) 38 cm
Material Polypropylene
Type Carrier Tray
Width (Metric) 24 cm
Length (English) 15 in.
Height (English) 4.5 in.
Shape Rectangular
Width (English) 9.5 in.
Autoclavable Autoclavable
Show More Show Less