missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ USB™ CycleSeq™ Thermostable DNA Polymerase

Catalog No. 792001000UN
Encompass
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
792001000UN 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 792001000UN Supplier Applied Biosystems™ Supplier No. 792001000UN

Includes

CycleSeq Thermostable DNA Polymerase 1,000 units, 1mL CycleSeq Dilution Buffer, 1mL 10X CycleSeq Reaction Buffer

USB™ CycleSeq™ Thermostable DNA Polymerase is a thermostable, DNA polymerase that is exonuclease-free and incorporates ddNTPs with a marked level of efficiency over standard thermostable DNA polymerases.

Specifications

Content And Storage CycleSeq Thermostable DNA Polymerase 1,000 units
• 1 mL CycleSeq Dilution Buffer
• 1 mL 10X CycleSeq Reaction Buffer

For Use With (Application) DNA Sequencing
Polymerase Thermostable DNA Polymerase
Quantity 1000 U

For Research Use Only. Not for use in diagnostic procedures.