missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine 98.0+%, TCI America™
Supplier: TCI America U002025G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C9H12N2O6 | |
| MFCD00006526 | |
| uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd | |
| 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
Specifications
| Uridine | |
| 165°C | |
| C9H12N2O6 | |
| MFCD00006526 | |
| DRTQHJPVMGBUCF-DHRBYNEYSA-N | |
| 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
| 89495556 | |
| ≥98.0% (HPLC,T) |
| 58-96-8 | |
| White | |
| 25 g | |
| uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd | |
| C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O | |
| 244.203 | |
| 244.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0942
RTECSNumber : YR1450000
TSCA : Yes