missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine, 99%

Uridine, 99%, C9H12N2O6, CAS Number-58-96-8, uracil, 1-beta-d-ribofuranosyl, uracil riboside, d-uridine, beta-uridine, 1-beta-d-ribofuranosyluracil, uridin, unii-whi7hq7h85, urd, b-uridine, uridine, 100g, 754904, 200-407-5, 244.2, DRTQHJPVMGBUCF-DHRBYNEYSA-N | CAS: 58-96-8 | C9H12N2O6 | 244.203 g/mol

Supplier:  Thermo Scientific Chemicals A1522714

Quantity 25 g

 View more versions of this product

Catalog No. AAA1522714


Only null left

Description

Description

Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.

Solubility
Soluble in water, dimethylsulfoxide, and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

58-96-8
244.203
DRTQHJPVMGBUCF-DHRBYNEYSA-N
89495556
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
C9H12N2O6
MFCD00006526
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione

Specifications

Uridine
166°C to 169°C
C9H12N2O6
MFCD00006526
149877
Soluble in water,dimethylsulfoxide,and methanol.
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
244.203
244.2
58-96-8
0.99
25 g
754904
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
DRTQHJPVMGBUCF-DHRBYNEYSA-N
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
89495556
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-407-5

RTECSNumber : YR1450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions