missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ uPSEM 792 hydrochloride

Catalog No. 68655
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68655 5 mg
686525 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68655 Supplier Tocris Bioscience™ Supplier No. 6865/5
Only null left

Highly potent PSAM4-GlyR and PSAM4-5-HT3 chimeric ion channel agonist; brain-penetrant

uPSEM 792 hydrochloride is a ultrapotent PSEM (uPSEM) agonist for PSAM4-GlyR and PSAM4-5HT3 (Ki = 0.7 nM for PSAM4-GlyR and <10 nM for PSAM4-5HT3). Exhibits >10,000-fold agonist selectivity for PSAM4-GlyR over α7-GlyR, α7-5HT3, and 5HT3-R, and 230-fold selectivity over α4β2 nAChR. Also weak partial agonist (∽10 %) at α4β2 nAChR. Retains the potency of varenicline (Cat.No. 3754) for PSAM4-GlyR with enhanced chemogenetic selectivity. Does not act as a substrate for P-glycoprotein pumps. Silences neurons in vivo. Brain-penetrant.

Specifications

Chemical Name or Material 1-Methyl-7,8,9,10-tetrahydro-1 H-6,10-methanoazepino[4,5-g]quinoxalin-2(6 H)-one hydrochloride
CAS 2341841-08-3
Quantity 5 mg
Target Pharmacologically Selective Effector Molecules/PSEM
Molecular Formula C14H15N3O.HCl
Formula Weight 277.75
Purity 98%