Learn More
Idex VHP Union Assembly
Zero dead volume unions ensure accurate analysis
$96.36 - $133.68
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | Swept Volume | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | Swept Volume | Price | Quantity & Availability | ||||
29008260
|
Idex
UP U 402 |
0.5 mm | 0.134 μL |
Each for $96.36
|
|
||||
05700234
|
Idex
UP U 412/DUP |
0.78 cm | 0.103 μL |
Each for $133.68
|
|
||||
05700247
|
Idex
UP U 435/DUP |
0.25 mm | 0.025 μL |
Each for $131.74
|
|
||||
Description
- Precision machined from 316 stainless steel to withstand high pressures
- Securely join fluid pathway components Holds tight connection even through vibration
- VHP ZDV union assembly
- 10-32 coned for 1/16 in. OD tubing
- Max pressure 20,000 psi (1380 bar)
Specifications
| Union | |
| 10000 psi | |
| Stainless Steel |