Learn More
Idex VHP Union Assembly
Zero dead volume unions ensure accurate analysis
Supplier: Idex UP U 412/DUP
Description
- Precision machined from 316 stainless steel to withstand high pressures
- Securely join fluid pathway components Holds tight connection even through vibration
- VHP ZDV union assembly
- 10-32 coned for 1/16 in. OD tubing
- Max pressure 20,000 psi (1380 bar)
Chemical Identifiers
| Union | |
| 10000 psi | |
| Stainless Steel |
Specifications
| Union | |
| 1.02 in. | |
| 0.103 μL | |
| 0.01 mm |
| 0.78 cm | |
| 10000 psi | |
| Stainless Steel |