Learn More
Idex ZDW Tefzel Low-Pressure Unions DFS Item
Make solid connections in low-pressure applications
$15.40 - $55.47
Specifications
Type Union
Temperature (Metric) Max 80°C
Material ETFE
Color Natural
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
05-700-152 DFS Item
View Documents
Idex
UPP623
Not Applicable
Each for $15.40
Only null left
05-700-159 DFS Item
View Documents
Idex
UPP630
w/2 P-200/P-245; 1/16 in.
Each for $36.58
Only null left
05-700-160 DFS Item
View Documents
Idex
UPP631
1/8 in. OD Tubing
Each for $36.58
Only null left
05-703-14 DFS Item
View Documents
Idex
UPP710
w/Tubing up to 3.17 mm O.D.
Each for $55.47
Only null left
Description
Description
Make solid connections in low-pressure applications
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
  • True ZDV union
  • ETFE body
Specifications
Specifications
Union
80°C
ETFE
Natural
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'