Learn More
Idex ZDW Tefzel Low-Pressure Unions DFS Item
Make solid connections in low-pressure applications
Supplier:  Idex UPP631
 View more versions of this product
Catalog No. 05-700-160
Only null left
Explore available promotions
Description
Description
Make solid connections in low-pressure applications
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
  • True ZDV union
  • ETFE body
Specifications
Specifications
Union
ETFE
1/8 in. OD Tubing
80°C
Natural
Product Suggestions
Product Suggestions