missing translation for 'onlineSavingsMsg'
Learn More

Idex Tefzel Low-Pressure Unions

Combine, split, or intersect fluid streams throughout your system

$106.57 - $111.36

Specifications

Type Tee
Diameter (English) Inner 0.05 in.
Pressure Rating 1000 psi
Material ETFE
Color Natural
View More Specs

Products 2
Catalog Number Mfr. No. Swept Volume Price Quantity & Availability  
Catalog Number Mfr. No. Swept Volume Price Quantity & Availability  
05700162
Encompass
View Documents Promotion Details
Idex
UPP 633
17.5 μL
Each for $106.57
Only null left
 
05700161
Encompass
View Documents Promotion Details
Idex
UP P 632
2.9 μL
Each for $111.36
Only null left
 
Description

Description

  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports

Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors. Tee connector assemblies come complete with nuts and ferrules.

Specifications

Specifications

Tee
1000 psi
Natural
0.05 in.
ETFE
Videos
SDS
Documents

Documents