Learn More
Idex Tefzel Low-Pressure Unions DFS Item
Combine, split, or intersect fluid streams throughout your system
Supplier:  Idex UPP632
 View more versions of this product
Catalog No. 05-700-161
Only null left
Description
Description
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors. Tee connector assemblies come complete with nuts and ferrules.
Specifications
Specifications
Tee
1000 psi
ETFE
0.05 in.
2.9 μL
Natural
Product Suggestions
Product Suggestions