Learn More
Idex Back Pressure Regulators and Accessories DFS Item
Placing back pressure regulator (usually 500 - 1,000 psi) between pump and injector can enhance pump's performance
$244.19 - $481.49
Specifications
Type Back Pressure Regulator Assembly
View More Specs
Products 9
Catalog Number Mfr. No. Material Max. Pressure Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Material Max. Pressure Volume (Metric) Price Quantity  
05-704-215 DFS Item
View Documents
Idex
UPP785
PEEK 40 psi 131 μL
Each for $357.64
Only null left
05-704-216 DFS Item
View Documents
Idex
UPP786
PEEK 75 psi 131 μL
Each for $384.45
Only null left
05-704-217 DFS Item
View Documents
Idex
UPP787
PEEK 100 psi 131 μL
Each for $357.64
Only null left
05-704-218 DFS Item
View Documents
Idex
UPP788
PEEK 250 psi 109 μL
Each for $354.25
Only null left
29-008-1219 DFS Item
View Documents
Idex
UPP765
PEEK 500 psi (34 bar) 89 μL
Each for $244.19
Only null left
29-008-1210 DFS Item
View Documents
Idex
UPU605
Stainless Steel 40 psi 129 μL
Each for $481.49
Only null left
29-008-1214 DFS Item
View Documents
Idex
UPU609
Stainless Steel 500 psi 129 μL
Each for $481.49
Only null left
29-008-1382 DFS Item
View Documents
Idex
UPP790
PEEK 5 psi 134 μL
Each for $357.64
Only null left
29-008-1383 DFS Item
View Documents
Idex
UPP791
PEEK 20 psi 134 μL
Each for $357.64
Only null left
Description
Description
Placing back pressure regulator (usually 500 - 1,000 psi) between pump and injector can enhance pump's performance
Bubbles enter the detector's flow cell, they can cause erratic baseline readings and loss of sensitivity. By placing a back pressure regulator (BPR) after the detector, the fluid remains under sufficient compression after leaving the column to prevent most bubbles from forming and thus helping to ensure a smoother baseline and increased sensitivity.
  • Standard versions, low-volume versions, and adjustable version available
  • Flow-independent pump preload for greater pump efficiency
  • Proven out-gassing protection
  • Multiple pressure ratings
  • 2,000 to 5,000psi (138 to 345 bar)
  • Biocompatible/chemically resistant materials
  • Only 6μLmaximum internal swept volume
  • Designed for multi-detector applications
  • 100% Biocompatible, inert materials
  • Replacement cartridges and holders sold separately
Specifications
Specifications
Back Pressure Regulator Assembly
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'