missing translation for 'onlineSavingsMsg'
Learn More

Idex Back Pressure Regulators and Accessories DFS Item

Placing back pressure regulator (usually 500 - 1,000 psi) between pump and injector can enhance pump's performance

Supplier:  Idex UPP791

 View more versions of this product

Catalog No. 29-008-1383


Only null left
Explore available promotions

Description

Description

Placing back pressure regulator (usually 500 - 1,000 psi) between pump and injector can enhance pump's performance

Bubbles enter the detector's flow cell, they can cause erratic baseline readings and loss of sensitivity. By placing a back pressure regulator (BPR) after the detector, the fluid remains under sufficient compression after leaving the column to prevent most bubbles from forming and thus helping to ensure a smoother baseline and increased sensitivity.

  • Standard versions, low-volume versions, and adjustable version available
  • Flow-independent pump preload for greater pump efficiency
  • Proven out-gassing protection
  • Multiple pressure ratings
  • 2,000 to 5,000psi (138 to 345 bar)
  • Biocompatible/chemically resistant materials
  • Only 6μLmaximum internal swept volume
  • Designed for multi-detector applications
  • 100% Biocompatible, inert materials
  • Replacement cartridges and holders sold separately

Specifications

Specifications

PEEK
20 psi
Back Pressure Regulator Assembly
134 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions