Learn More
MilliporeSigma™ Supelco™ Untreated Fused Silica Tubing
Supplier: MilliporeSigma™ Supelco™ 24061
Chemical Identifiers
| Tubing | |
| Untreated | |
| Gas Chromatography (GC) | |
| Fused Silica |
Specifications
| Tubing | |
| Gas Chromatography (GC) | |
| 60 m |
| Untreated | |
| 0.25 mm | |
| Fused Silica |